The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017629.3 | NP_060099.2 | 3 - 4 | 497 | 67 |
XM_005270578.3 | XP_005270635.1 | 3 - 4 | 382 | 67 | |
XM_005270579.3 | XP_005270636.1 | 2 - 3 | 190 | 67 | |
XM_017000529.1 | XP_016856018.1 | 3 - 4 | 394 | 67 | |
GenBank mRNA | AB046787.1 | - | 3 - 4 | 497 | 67 |
AL831908.1 | - | 2 - 3 | 284 | 67 | |
BC152450.1 | - | 3 - 4 | 497 | 67 |