The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017629.3 | NP_060099.2 | 8 - 9 | 1186 | 69 |
XM_005270578.3 | XP_005270635.1 | 8 - 9 | 1071 | 69 | |
XM_005270579.3 | XP_005270636.1 | 7 - 8 | 879 | 69 | |
XM_011540883.2 | XP_011539185.1 | 5 - 6 | 675 | 69 | |
XM_011540884.2 | XP_011539186.1 | 5 - 6 | 538 | 69 | |
XM_017000529.1 | XP_016856018.1 | 8 - 9 | 1083 | 69 | |
GenBank mRNA | AB046787.1 | - | 8 - 9 | 1186 | 69 |
AL831908.1 | - | 7 - 8 | 973 | 69 | |
BC152450.1 | - | 8 - 9 | 1186 | 69 |