The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201472.1 | NP_001188401.1 | 27 - 28 | 3156 | 153 |
NM_001201473.1 | NP_001188402.1 | 25 - 26 | 2955 | 153 | |
NM_024535.4 | NP_078811.3 | 27 - 28 | 3210 | 153 | |
GenBank mRNA | AK025674.1 | - | 27 - 28 | 2819 | 153 |
AK094596.1 | - | 27 - 28 | 2829 | 153 | |
AK294045.1 | - | 27 - 28 | 2791 | 153 | |
AK296723.1 | - | 27 - 28 | 2788 | 153 | |
AK296807.1 | - | 25 - 26 | 2621 | 153 | |
BC117289.1 | - | 27 - 28 | 2829 | 153 | |
BC117291.1 | - | 27 - 28 | 2829 | 153 |