The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167733.2 | NP_001161205.1 | 18 - 19 | 1745 | 88 |
NM_001167734.1 | NP_001161206.1 | 19 - 20 | 1980 | 88 | |
NM_020442.5 | NP_065175.4 | 19 - 20 | 1973 | 88 | |
GenBank mRNA | AB067472.1 | - | 18 - 19 | 2441 | 88 |
AK000511.1 | - | 18 - 19 | 1650 | 88 | |
AK025618.1 | - | 7 - 8 | 565 | 88 | |
AK125069.1 | - | 8 - 9 | 1728 | 88 | |
AK293780.1 | - | 18 - 19 | 1734 | 88 | |
AK303415.1 | - | 19 - 20 | 1962 | 88 | |
AK304807.1 | - | 19 - 20 | 1980 | 88 | |
AK315733.1 | - | 8 - 9 | 731 | 88 | |
BC008844.2 | - | 2 - 3 | 168 | 88 | |
BC009355.2 | - | 9 - 10 | 821 | 88 | |
BC073838.1 | - | 7 - 8 | 591 | 88 | |
BC112054.1 | - | 18 - 19 | 1835 | 88 | |
BC113605.1 | - | 18 - 19 | 1835 | 88 | |
BC143535.1 | - | 18 - 19 | 1834 | 88 | |
BC143536.1 | - | 18 - 19 | 1828 | 88 |