Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTRA4
Entrez Gene ID
203100
Gene Name
HtrA serine peptidase 4
Gene Aliases
-
Chromosome Location
Chr.8: 38974149 - 38988662 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153692.3 NP_710159.1 5 - 6 1115 158
XM_011544431.2 XP_011542733.1 5 - 6 1115 158
GenBank mRNA AK075205.1 - 5 - 6 1086 158
AK300197.1 - 5 - 6 994 158
BC057765.1 - 5 - 6 1047 158

View Full Product Details