The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024830.3 | NP_079106.3 | 5 - 6 | 791 | 113 |
XM_005248373.3 | XP_005248430.1 | 6 - 7 | 699 | 113 | |
XM_011514132.1 | XP_011512434.1 | 5 - 6 | 800 | 113 | |
XM_011514134.1 | XP_011512436.1 | 5 - 6 | 566 | 113 | |
GenBank mRNA | AB244719.2 | - | 5 - 6 | 674 | 113 |
AK090444.1 | - | 3 - 4 | 270 | 113 | |
BC020166.1 | - | 3 - 4 | 257 | 113 |