Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP6R3
Entrez Gene ID
55291
Gene Name
protein phosphatase 6 regulatory subunit 3
Gene Aliases
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3
Chromosome Location
Chr.11: 68460712 - 68615334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164160.1 NP_001157632.1 8 - 9 1106 88
NM_001164161.1 NP_001157633.1 8 - 9 1106 88
NM_001164162.1 NP_001157634.1 8 - 9 1106 88
NM_001164163.1 NP_001157635.1 8 - 9 1106 88
NM_001164164.1 NP_001157636.1 9 - 10 1211 88
NM_018312.4 NP_060782.2 9 - 10 1211 88
XM_006718608.3 XP_006718671.1 9 - 10 1222 88
XM_006718612.3 XP_006718675.1 9 - 10 1222 88
XM_006718613.2 XP_006718676.1 9 - 10 1222 88
XM_006718614.2 XP_006718677.1 9 - 10 1222 88
XM_006718618.3 XP_006718681.1 9 - 10 1222 88
XM_006718621.2 XP_006718684.1 9 - 10 1222 88
XM_006718622.3 XP_006718685.1 9 - 10 1222 88
XM_006718623.3 XP_006718686.1 9 - 10 1205 88
XM_006718624.2 XP_006718687.1 9 - 10 1222 88
XM_006718627.3 XP_006718690.1 9 - 10 1222 88
XM_011545135.1 XP_011543437.1 9 - 10 1222 88
XM_011545136.2 XP_011543438.1 8 - 9 1105 88
XM_011545137.2 XP_011543439.1 9 - 10 1205 88
XM_011545138.2 XP_011543440.1 10 - 11 1423 88
XM_011545139.1 XP_011543441.1 9 - 10 1222 88
XM_011545140.2 XP_011543442.1 9 - 10 1222 88
XM_011545141.1 XP_011543443.1 9 - 10 1222 88
XM_011545142.2 XP_011543444.1 9 - 10 1222 88
XM_011545143.1 XP_011543445.1 9 - 10 1222 88
XM_011545145.2 XP_011543447.1 9 - 10 1222 88
XM_017017962.1 XP_016873451.1 8 - 9 1060 88
XM_017017963.1 XP_016873452.1 9 - 10 1146 88
XM_017017964.1 XP_016873453.1 9 - 10 1222 88
XM_017017965.1 XP_016873454.1 8 - 9 1105 88
XM_017017966.1 XP_016873455.1 9 - 10 1308 88
XM_017017967.1 XP_016873456.1 9 - 10 1222 88
XM_017017968.1 XP_016873457.1 8 - 9 1105 88
XM_017017969.1 XP_016873458.1 9 - 10 1222 88
XM_017017970.1 XP_016873459.1 8 - 9 1105 88
XM_017017971.1 XP_016873460.1 9 - 10 1222 88
XM_017017972.1 XP_016873461.1 8 - 9 1078 88
XM_017017973.1 XP_016873462.1 9 - 10 1205 88
XM_017017974.1 XP_016873463.1 9 - 10 1222 88
XM_017017975.1 XP_016873464.1 9 - 10 1222 88
XM_017017976.1 XP_016873465.1 8 - 9 1105 88
XM_017017977.1 XP_016873466.1 8 - 9 993 88
XM_017017978.1 XP_016873467.1 7 - 8 844 88
XM_017017979.1 XP_016873468.1 7 - 8 844 88
XM_017017980.1 XP_016873469.1 7 - 8 844 88
XM_017017981.1 XP_016873470.1 7 - 8 844 88
XM_017017982.1 XP_016873471.1 7 - 8 844 88
XM_017017983.1 XP_016873472.1 8 - 9 1243 88
XM_017017984.1 XP_016873473.1 8 - 9 1173 88
GenBank mRNA AB046778.2 - 8 - 9 1054 88
AF264779.1 - 8 - 9 1116 88
AF264780.1 - 8 - 9 1116 88
AK057250.1 - 2 - 3 179 88
AK315136.1 - 8 - 9 1075 88
BC105933.1 - 7 - 8 974 88
BC105934.1 - 7 - 8 974 88
BC107599.1 - 7 - 8 974 88
CR933658.1 - 8 - 9 1069 88
DQ111954.2 - 6 - 7 839 88

View Full Product Details