Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TARSL2
Entrez Gene ID
123283
Gene Name
threonyl-tRNA synthetase like 2
Gene Aliases
-
Chromosome Location
Chr.15: 101653752 - 101724604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152334.2 NP_689547.2 1 - 2 353 103
XM_006720386.3 XP_006720449.1 1 - 2 515 103
XM_017021912.1 XP_016877401.1 1 - 2 515 103
GenBank mRNA AK056653.1 - 1 - 2 103
BC132671.1 - 1 - 2 321 103
BC136315.1 - 1 - 2 321 103
BC143943.1 - 1 - 2 321 103

View Full Product Details