Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TARSL2
Entrez Gene ID
123283
Gene Name
threonyl-tRNA synthetase like 2
Gene Aliases
-
Chromosome Location
Chr.15: 101653752 - 101724604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152334.2 NP_689547.2 2 - 3 425 145
XM_006720386.3 XP_006720449.1 2 - 3 587 145
XM_017021912.1 XP_016877401.1 2 - 3 587 145
GenBank mRNA AK056653.1 - 2 - 3 415 145
AL833188.1 - 2 - 3 91 145
BC132671.1 - 2 - 3 393 145
BC136315.1 - 2 - 3 393 145
BC143943.1 - 2 - 3 393 145

View Full Product Details