Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
Chromosome Location
Chr.11: 74696428 - 74731385 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278473.2 NP_001265402.1 8 - 9 1446 72
NM_001304390.1 NP_001291319.1 7 - 8 1338 72
NM_001304416.1 NP_001291345.1 6 - 7 1244 72
NM_001304417.1 NP_001291346.1 5 - 6 1036 72
NM_015424.5 NP_056239.3 8 - 9 1446 72
GenBank mRNA AF332891.1 - 5 - 6 727 72
AK304519.1 - 8 - 9 1214 72
AL110168.1 - 2 - 3 348 72
AY163868.1 - 8 - 9 952 72
AY279090.1 - 8 - 9 1243 72
AY279092.1 - 6 - 7 1244 72
AY279093.1 - 6 - 7 1244 72
AY279094.1 - 5 - 6 1150 72
AY358522.1 - 8 - 9 1235 72
BC142623.1 - 8 - 9 1446 72

View Full Product Details