The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185106.1 | NP_001172035.1 | 2 - 3 | 1194 | 63 |
NM_153268.3 | NP_695000.1 | 2 - 3 | 1194 | 63 | |
GenBank mRNA | AK056141.1 | - | 2 - 3 | 1194 | 63 |
BC121155.1 | - | 2 - 3 | 948 | 63 | |
BC121156.1 | - | 2 - 3 | 948 | 63 |