Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCN8
Entrez Gene ID
138307
Gene Name
lipocalin 8
Gene Aliases
EP17, LCN5
Chromosome Location
Chr.9: 136754388 - 136758551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178469.3 NP_848564.2 6 - 7 800 104
GenBank mRNA AK126902.1 - 5 - 6 2353 104
BC130465.1 - 6 - 7 755 104
BC132714.1 - 6 - 7 755 104

View Full Product Details