The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146076.1 | NP_001139548.1 | 10 - 11 | 1148 | 121 |
NM_024604.2 | NP_078880.2 | 11 - 12 | 1307 | 121 | |
XM_011538741.2 | XP_011537043.1 | 9 - 10 | 1058 | 121 | |
GenBank mRNA | AK025561.1 | - | 11 - 12 | 1237 | 121 |
AK299465.1 | - | 10 - 11 | 1113 | 121 |