The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282116.1 | NP_001269045.1 | 12 - 13 | 1719 | 81 |
NM_002919.3 | NP_002910.1 | 11 - 12 | 1464 | 81 | |
NM_134428.2 | NP_602304.1 | 13 - 14 | 1786 | 81 | |
XM_006716840.1 | XP_006716903.1 | 12 - 13 | 1632 | 81 | |
XM_006716843.1 | XP_006716906.1 | 13 - 14 | 1632 | 81 | |
XM_006716844.1 | XP_006716907.1 | 13 - 14 | 1648 | 81 | |
XM_006716845.2 | XP_006716908.1 | 12 - 13 | 1710 | 81 | |
XM_006716847.3 | XP_006716910.1 | 7 - 8 | 765 | 81 | |
XM_011517998.1 | XP_011516300.1 | 13 - 14 | 1700 | 81 | |
XM_011517999.1 | XP_011516301.1 | 13 - 14 | 1664 | 81 | |
XM_011518000.1 | XP_011516302.1 | 11 - 12 | 1635 | 81 | |
XM_011518001.1 | XP_011516303.1 | 14 - 15 | 1714 | 81 | |
XM_017015000.1 | XP_016870489.1 | 13 - 14 | 1651 | 81 | |
XM_017015001.1 | XP_016870490.1 | 10 - 11 | 1389 | 81 | |
XM_017015002.1 | XP_016870491.1 | 6 - 7 | 3171 | 81 | |
GenBank mRNA | AK289540.1 | - | 12 - 13 | 1517 | 81 |
BC022191.2 | - | 13 - 14 | 1767 | 81 | |
X76092.1 | - | 11 - 12 | 1464 | 81 |