The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306190.1 | NP_001293119.1 | 5 - 6 | 650 | 64 |
NM_001698.2 | NP_001689.1 | 6 - 7 | 692 | 64 | |
XM_005252066.3 | XP_005252123.1 | 7 - 8 | 735 | 64 | |
XM_005252067.4 | XP_005252124.1 | 7 - 8 | 737 | 64 | |
XM_005252069.4 | XP_005252126.1 | 7 - 8 | 738 | 64 | |
XM_006717150.3 | XP_006717213.1 | 6 - 7 | 651 | 64 | |
XM_011518802.2 | XP_011517104.1 | 7 - 8 | 761 | 64 | |
XM_017014849.1 | XP_016870338.1 | 6 - 7 | 692 | 64 | |
XM_017014850.1 | XP_016870339.1 | 7 - 8 | 832 | 64 | |
XM_017014851.1 | XP_016870340.1 | 7 - 8 | 761 | 64 | |
GenBank mRNA | BC020722.1 | - | 5 - 6 | 594 | 64 |
X79888.1 | - | 6 - 7 | 661 | 64 |