Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPECC1
Entrez Gene ID
92521
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases
CYTSB, HCMOGT-1, HCMOGT1, NSP
Chromosome Location
Chr.17: 20009301 - 20319026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033553.2 NP_001028725.1 8 - 9 2550 121
NM_001033555.2 NP_001028727.1 6 - 7 2459 121
NM_001243439.1 NP_001230368.1 8 - 9 2609 121
XM_005256860.2 XP_005256917.1 5 - 6 879 121
XM_017025317.1 XP_016880806.1 7 - 8 2526 121
XM_017025318.1 XP_016880807.1 7 - 8 2526 121
XM_017025319.1 XP_016880808.1 8 - 9 2668 121
XM_017025320.1 XP_016880809.1 8 - 9 2644 121
XM_017025321.1 XP_016880810.1 6 - 7 2483 121
GenBank mRNA AK295093.1 - 8 - 9 2582 121
AK295811.1 - 5 - 6 2086 121
AK298601.1 - 5 - 6 1291 121
AK301318.1 - 5 - 6 801 121
AK304186.1 - 6 - 7 2381 121
BC021123.1 - 6 - 7 2357 121

View Full Product Details