Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGAP2
- Entrez Gene ID
-
27315
- Gene Name
- post-GPI attachment to proteins 2
- Gene Aliases
-
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
- Chromosome Location
-
Chr.11: 3797724 - 3826371 on Build GRCh38
View Full Product Details