The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016320.4 | NP_057404.2 | 20 - 21 | 3164 | 81 |
NM_139132.3 | NP_624358.2 | 20 - 21 | 3164 | 81 | |
GenBank mRNA | AF071076.1 | - | 20 - 21 | 2867 | 81 |
AF071077.1 | - | 20 - 21 | 2867 | 81 | |
AF231130.1 | - | 20 - 21 | 2887 | 81 |