The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005387.6 | NP_005378.4 | 4 - 5 | 777 | 82 |
NM_016320.4 | NP_057404.2 | 4 - 5 | 777 | 82 | |
NM_139131.4 | NP_624357.1 | 4 - 5 | 777 | 82 | |
NM_139132.3 | NP_624358.2 | 4 - 5 | 777 | 82 | |
GenBank mRNA | AB040538.1 | - | 4 - 5 | 532 | 82 |
AF071076.1 | - | 4 - 5 | 480 | 82 | |
AF071077.1 | - | 4 - 5 | 480 | 82 | |
AF231130.1 | - | 4 - 5 | 500 | 82 | |
AK292882.1 | - | 4 - 5 | 517 | 82 | |
BC041136.1 | - | 4 - 5 | 526 | 82 | |
BG773331.1 | - | 4 - 5 | 700 | 82 | |
KU847959.1 | - | - | 356 | 82 | |
U41815.1 | - | 4 - 5 | 500 | 82 |