The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127325.1 | NP_001120797.1 | 1 - 2 | 207 | 105 |
GenBank mRNA | BI517654.1 | - | 1 - 2 | 207 | 105 |