The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000368.4 | NP_000359.1 | 12 - 13 | 1498 | 92 |
NM_001162426.1 | NP_001155898.1 | 12 - 13 | 1495 | 92 | |
NM_001162427.1 | NP_001155899.1 | 11 - 12 | 1345 | 92 | |
XM_005272211.1 | XP_005272268.1 | 12 - 13 | 1543 | 92 | |
XM_006717271.1 | XP_006717334.1 | 12 - 13 | 1478 | 92 | |
XM_011518979.2 | XP_011517281.1 | 12 - 13 | 1492 | 92 | |
XM_017015096.1 | XP_016870585.1 | 13 - 14 | 1957 | 92 | |
XM_017015097.1 | XP_016870586.1 | 13 - 14 | 1928 | 92 | |
XM_017015098.1 | XP_016870587.1 | 12 - 13 | 1540 | 92 | |
XM_017015099.1 | XP_016870588.1 | 11 - 12 | 1636 | 92 | |
XM_017015100.1 | XP_016870589.1 | 11 - 12 | 1607 | 92 | |
XM_017015101.1 | XP_016870590.1 | 11 - 12 | 1634 | 92 | |
GenBank mRNA | AF013168.1 | - | 12 - 13 | 1485 | 92 |
AK303030.1 | - | 11 - 12 | 1328 | 92 | |
AK308412.1 | - | 12 - 13 | 1478 | 92 | |
BC108668.1 | - | 12 - 13 | 1485 | 92 | |
KJ534953.1 | - | 10 - 11 | 1264 | 92 |