The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000368.4 | NP_000359.1 | 20 - 21 | 2857 | 65 |
NM_001162426.1 | NP_001155898.1 | 20 - 21 | 2854 | 65 | |
NM_001162427.1 | NP_001155899.1 | 19 - 20 | 2704 | 65 | |
XM_005272211.1 | XP_005272268.1 | 20 - 21 | 2902 | 65 | |
XM_006717271.1 | XP_006717334.1 | 20 - 21 | 2837 | 65 | |
XM_011518979.2 | XP_011517281.1 | 20 - 21 | 2851 | 65 | |
XM_017015096.1 | XP_016870585.1 | 21 - 22 | 3316 | 65 | |
XM_017015097.1 | XP_016870586.1 | 21 - 22 | 3287 | 65 | |
XM_017015098.1 | XP_016870587.1 | 20 - 21 | 2899 | 65 | |
XM_017015099.1 | XP_016870588.1 | 19 - 20 | 2995 | 65 | |
XM_017015100.1 | XP_016870589.1 | 19 - 20 | 2966 | 65 | |
XM_017015101.1 | XP_016870590.1 | 19 - 20 | 2993 | 65 | |
GenBank mRNA | AF013168.1 | - | 20 - 21 | 2844 | 65 |
AK303030.1 | - | 19 - 20 | 2687 | 65 | |
D87683.1 | - | 7 - 8 | 1228 | 65 | |
KJ534953.1 | - | 18 - 19 | 2623 | 65 |