Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOP2B
Entrez Gene ID
7155
Gene Name
topoisomerase (DNA) II beta
Gene Aliases
TOPIIB, top2beta
Chromosome Location
Chr.3: 25597905 - 25664936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001068.3 NP_001059.2 24 - 25 3315 140
XM_005265427.3 XP_005265484.1 24 - 25 3894 140
XM_011534057.2 XP_011532359.1 24 - 25 3304 140
GenBank mRNA AB208879.1 - 21 - 22 2873 140
AK296510.1 - 25 - 26 3370 140
AK307450.1 - 26 - 27 3315 140
X53662.1 - 1 - 2 129 140
X68060.1 - 24 - 25 3240 140
Z15115.1 - 10 - 11 1470 140

View Full Product Details