The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172364.4 | NP_758952.4 | 29 - 30 | 2948 | 80 |
XM_011521041.2 | XP_011519343.1 | 28 - 29 | 6787 | 80 | |
GenBank mRNA | AF516695.1 | - | 29 - 30 | 3080 | 80 |
AK301234.1 | - | 2 - 3 | 189 | 80 | |
BC150186.1 | - | 28 - 29 | 2871 | 80 | |
BI908421.1 | - | 2 - 3 | 155 | 80 | |
BX537436.1 | - | 29 - 30 | 2911 | 80 | |
BX537437.1 | - | 29 - 30 | 2949 | 80 |