The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015633.2 | NP_056448.1 | 5 - 6 | 867 | 68 |
GenBank mRNA | AK001534.1 | - | 5 - 6 | 856 | 68 |