The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144772.1 | NP_001138244.1 | 11 - 12 | 911 | 90 |
NM_003580.3 | NP_003571.2 | 11 - 12 | 971 | 90 | |
GenBank mRNA | AK292679.1 | - | 11 - 12 | 851 | 90 |
AK294009.1 | - | 11 - 12 | 911 | 90 | |
AK313991.1 | - | 11 - 12 | 882 | 90 | |
BC041124.1 | - | 11 - 12 | 953 | 90 | |
X96586.1 | - | 11 - 12 | 769 | 90 |