Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP63
Entrez Gene ID
80254
Gene Name
centrosomal protein 63
Gene Aliases
SCKL6
Chromosome Location
Chr.3: 134485733 - 134629607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042383.1 NP_001035842.1 3 - 4 569 98
NM_001042384.1 NP_001035843.1 3 - 4 569 98
NM_001042400.1 NP_001035859.1 3 - 4 397 98
NM_025180.3 NP_079456.2 4 - 5 673 98
XM_005247795.4 XP_005247852.1 3 - 4 346 98
XM_005247797.3 XP_005247854.1 3 - 4 601 98
XM_005247799.3 XP_005247856.1 3 - 4 396 98
XM_006713760.3 XP_006713823.1 3 - 4 278 98
XM_011513194.2 XP_011511496.1 3 - 4 395 98
XM_017007244.1 XP_016862733.1 3 - 4 389 98
XM_017007245.1 XP_016862734.1 3 - 4 386 98
XM_017007246.1 XP_016862735.1 3 - 4 278 98
XM_017007247.1 XP_016862736.1 3 - 4 395 98
XM_017007248.1 XP_016862737.1 3 - 4 562 98
XM_017007249.1 XP_016862738.1 4 - 5 651 98
XM_017007250.1 XP_016862739.1 3 - 4 387 98
XM_017007251.1 XP_016862740.1 3 - 4 385 98
XM_017007252.1 XP_016862741.1 3 - 4 302 98
XM_017007253.1 XP_016862742.1 3 - 4 381 98
XM_017007254.1 XP_016862743.1 3 - 4 278 98
XM_017007255.1 XP_016862744.1 3 - 4 601 98
XM_017007256.1 XP_016862745.1 4 - 5 651 98
XM_017007257.1 XP_016862746.1 3 - 4 383 98
XM_017007258.1 XP_016862747.1 3 - 4 278 98
XM_017007259.1 XP_016862748.1 3 - 4 395 98
XM_017007260.1 XP_016862749.1 3 - 4 287 98
XM_017007261.1 XP_016862750.1 3 - 4 346 98
XM_017007262.1 XP_016862751.1 3 - 4 395 98
XM_017007263.1 XP_016862752.1 3 - 4 601 98
XM_017007264.1 XP_016862753.1 4 - 5 651 98
XM_017007265.1 XP_016862754.1 3 - 4 379 98
XM_017007266.1 XP_016862755.1 3 - 4 395 98
XM_017007267.1 XP_016862756.1 3 - 4 562 98
XM_017007268.1 XP_016862757.1 3 - 4 566 98
XM_017007269.1 XP_016862758.1 4 - 5 651 98
GenBank mRNA AK023448.1 - 3 - 4 343 98
AK023738.1 - 3 - 4 510 98
AK056465.1 - 4 - 5 651 98
BC014050.2 - 3 - 4 558 98
BC068997.1 - 3 - 4 317 98
BI561425.1 - 3 - 4 400 98
CX751767.1 - 3 - 4 509 98

View Full Product Details