Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
79446
Gene Name
WD repeat domain 25
Gene Aliases
C14orf67
Chromosome Location
Chr.14: 100376410 - 100530303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161476.1 NP_001154948.1 2 - 3 921 95
NM_024515.4 NP_078791.3 2 - 3 1052 95
XM_005268056.4 XP_005268113.1 2 - 3 928 95
XM_011537150.2 XP_011535452.1 2 - 3 1175 95
GenBank mRNA AK291960.1 - 2 - 3 884 95
AY295082.1 - 2 - 3 935 95

View Full Product Details