The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014974.2 | NP_055789.1 | 22 - 23 | 2880 | 73 |
XM_005252426.3 | XP_005252483.1 | 23 - 24 | 3039 | 73 | |
XM_005252427.3 | XP_005252484.1 | 22 - 23 | 2920 | 73 | |
XM_005252428.4 | XP_005252485.1 | 23 - 24 | 3090 | 73 | |
XM_005252430.2 | XP_005252487.2 | 6 - 7 | 1130 | 73 | |
XM_011519428.2 | XP_011517730.1 | 23 - 24 | 3021 | 73 | |
XM_011519429.2 | XP_011517731.1 | 23 - 24 | 3019 | 73 | |
XM_011519430.2 | XP_011517732.1 | 22 - 23 | 2896 | 73 | |
XM_011519431.2 | XP_011517733.1 | 22 - 23 | 2896 | 73 | |
XM_011519432.1 | XP_011517734.1 | 22 - 23 | 3321 | 73 | |
XM_011519433.2 | XP_011517735.1 | 6 - 7 | 2129 | 73 | |
XM_017015977.1 | XP_016871466.1 | 6 - 7 | 1141 | 73 | |
GenBank mRNA | AB023151.2 | - | 22 - 23 | 2878 | 73 |
AK126063.1 | - | 24 - 25 | 3133 | 73 | |
AK298353.1 | - | 6 - 7 | 977 | 73 | |
AK309715.1 | - | 6 - 7 | 892 | 73 | |
AK310018.1 | - | 5 - 6 | 1133 | 73 | |
BC063313.1 | - | 22 - 23 | 2880 | 73 |