The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024710.2 | NP_078986.1 | 3 - 4 | 521 | 64 |
GenBank mRNA | AK027122.1 | - | 3 - 4 | 484 | 64 |
CR457377.1 | - | 2 - 3 | 347 | 64 |