Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHID1
Entrez Gene ID
66005
Gene Name
chitinase domain containing 1
Gene Aliases
GL008, SI-CLP, SICLP
Chromosome Location
Chr.11: 867859 - 915058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142674.1 NP_001136146.1 9 - 10 949 75
NM_001142675.1 NP_001136147.1 8 - 9 1059 75
NM_001142676.1 NP_001136148.1 9 - 10 920 75
NM_023947.3 NP_076436.3 8 - 9 845 75
XM_005253072.2 XP_005253129.1 8 - 9 932 75
XM_006718282.2 XP_006718345.1 9 - 10 920 75
XM_011520299.1 XP_011518601.1 8 - 9 1156 75
XM_011520301.2 XP_011518603.1 8 - 9 1156 75
XM_011520302.2 XP_011518604.1 8 - 9 1156 75
XM_017018166.1 XP_016873655.1 9 - 10 1006 75
XM_017018167.1 XP_016873656.1 8 - 9 811 75
XM_017018168.1 XP_016873657.1 9 - 10 932 75
GenBank mRNA AF212229.1 - 9 - 10 883 75
AK027711.1 - 8 - 9 794 75
AK075413.1 - 9 - 10 856 75
AK124697.1 - 8 - 9 1421 75
AK225025.1 - 9 - 10 885 75
AK225398.1 - 9 - 10 868 75
AK297743.1 - 8 - 9 926 75
AK301245.1 - 8 - 9 905 75
BC000001.2 - 9 - 10 869 75
BC013642.1 - 6 - 7 481 75
BC095409.1 - 8 - 9 781 75

View Full Product Details