The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004815.3 | NP_004806.3 | 9 - 10 | 1240 | 101 |
XM_006711048.3 | XP_006711111.1 | 8 - 9 | 874 | 101 | |
XM_011542438.2 | XP_011540740.1 | 9 - 10 | 1115 | 101 | |
XM_011542439.2 | XP_011540741.1 | 9 - 10 | 960 | 101 | |
XM_011542440.2 | XP_011540742.1 | 8 - 9 | 834 | 101 | |
GenBank mRNA | AB208873.1 | - | 8 - 9 | 846 | 101 |
AK307733.1 | - | 9 - 10 | 1069 | 101 | |
AK310491.1 | - | 9 - 10 | 960 | 101 | |
BC022483.1 | - | 9 - 10 | 1122 | 101 | |
BC067839.1 | - | 9 - 10 | 1235 | 101 | |
BC093741.1 | - | 8 - 9 | 913 | 101 | |
BC093767.1 | - | 8 - 9 | 916 | 101 | |
BX508890.1 | - | 2 - 3 | 170 | 101 | |
U90920.1 | - | 9 - 10 | 1054 | 101 |