The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016148.3 | NP_057232.2 | 12 - 13 | 1885 | 78 |
XM_006723233.3 | XP_006723296.1 | 13 - 14 | 2283 | 78 | |
XM_011527013.2 | XP_011525315.1 | 13 - 14 | 2283 | 78 | |
XM_011527014.2 | XP_011525316.1 | 13 - 14 | 2283 | 78 | |
GenBank mRNA | AF163302.1 | - | 12 - 13 | 1885 | 78 |