Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHANK1
Entrez Gene ID
50944
Gene Name
SH3 and multiple ankyrin repeat domains 1
Gene Aliases
SPANK-1, SSTRIP, synamon
Chromosome Location
Chr.19: 50659257 - 50719779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016148.3 NP_057232.2 13 - 14 1958 58
XM_006723233.3 XP_006723296.1 14 - 15 2356 58
XM_011527013.2 XP_011525315.1 14 - 15 2356 58
GenBank mRNA AF163302.1 - 13 - 14 1958 58

View Full Product Details