The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016148.3 | NP_057232.2 | 18 - 19 | 2412 | 99 |
XM_006723233.3 | XP_006723296.1 | 19 - 20 | 2810 | 99 | |
XM_011527014.2 | XP_011525316.1 | 18 - 19 | 2783 | 99 | |
GenBank mRNA | AF163302.1 | - | 18 - 19 | 2412 | 99 |
AF226728.1 | - | 4 - 5 | 554 | 99 |