Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHANK1
Entrez Gene ID
50944
Gene Name
SH3 and multiple ankyrin repeat domains 1
Gene Aliases
SPANK-1, SSTRIP, synamon
Chromosome Location
Chr.19: 50659257 - 50719779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016148.3 NP_057232.2 1 - 2 278 90
XM_006723233.3 XP_006723296.1 2 - 3 676 90
XM_011527013.2 XP_011525315.1 2 - 3 676 90
XM_011527014.2 XP_011525316.1 2 - 3 676 90
GenBank mRNA AF163302.1 - 1 - 2 278 90

View Full Product Details