The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024537.3 | NP_078813.1 | 2 - 3 | 355 | 90 |
XM_006719953.2 | XP_006720016.1 | 2 - 3 | 435 | 90 | |
XM_011521116.1 | XP_011519418.1 | 2 - 3 | 147 | 90 | |
XM_011521117.2 | XP_011519419.1 | 2 - 3 | 291 | 90 | |
XM_011521118.2 | XP_011519420.1 | 2 - 3 | 291 | 90 | |
XM_017020742.1 | XP_016876231.1 | 2 - 3 | 291 | 90 | |
GenBank mRNA | AB062436.1 | - | 2 - 3 | 266 | 90 |
AK022180.1 | - | 2 - 3 | 291 | 90 | |
BC007220.1 | - | 2 - 3 | 278 | 90 | |
DA966145.1 | - | 2 - 3 | 310 | 90 | |
HY150557.1 | - | 2 - 3 | 356 | 90 | |
HY160154.1 | - | 2 - 3 | 337 | 90 |