The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024537.3 | NP_078813.1 | 9 - 10 | 1080 | 106 |
XM_006719953.2 | XP_006720016.1 | 9 - 10 | 1160 | 106 | |
XM_011521116.1 | XP_011519418.1 | 10 - 11 | 922 | 106 | |
XM_017020741.1 | XP_016876230.1 | 8 - 9 | 808 | 106 | |
GenBank mRNA | AB062436.1 | - | 9 - 10 | 997 | 106 |
AK022180.1 | - | 9 - 10 | 1016 | 106 | |
AK091992.1 | - | 2 - 3 | 90 | 106 | |
AK092947.1 | - | 2 - 3 | 303 | 106 | |
AK301907.1 | - | 8 - 9 | 1185 | 106 | |
BC007220.1 | - | 9 - 10 | 1003 | 106 | |
BX648882.1 | - | 1 - 2 | 3637 | 106 |