Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUFY1
Entrez Gene ID
80230
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
RABIP4, ZFYVE12
Chromosome Location
Chr.5: 179550551 - 179610026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040451.2 NP_001035541.1 12 - 13 1660 115
NM_001040452.2 NP_001035542.1 13 - 14 1471 115
NM_025158.4 NP_079434.3 13 - 14 1646 115
XM_005265993.4 XP_005266050.1 10 - 11 1017 115
XM_006714921.3 XP_006714984.2 12 - 13 1590 115
XM_006714922.3 XP_006714985.1 10 - 11 1450 115
XM_017009891.1 XP_016865380.1 12 - 13 1405 115
XM_017009892.1 XP_016865381.1 11 - 12 1443 115
XM_017009893.1 XP_016865382.1 10 - 11 1377 115
XM_017009894.1 XP_016865383.1 14 - 15 1612 115
XM_017009895.1 XP_016865384.1 13 - 14 1596 115
GenBank mRNA AB209507.1 - 14 - 15 1516 115
AF361055.2 - 13 - 14 1619 115
AK025904.1 - 7 - 8 709 115
AK075021.1 - 12 - 13 1560 115
AK093848.1 - 5 - 6 1578 115
AK291926.1 - 12 - 13 1660 115
BC032571.1 - 13 - 14 1390 115

View Full Product Details