The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309516.1 | NP_001296445.1 | 6 - 7 | 1017 | 106 |
NM_005048.3 | NP_005039.1 | 6 - 7 | 913 | 106 | |
GenBank mRNA | AK294181.1 | - | 6 - 7 | 1017 | 106 |
AY497546.1 | - | 6 - 7 | 699 | 106 | |
BC036811.1 | - | 6 - 7 | 912 | 106 | |
U25128.1 | - | 6 - 7 | 841 | 106 |