Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRG2
Entrez Gene ID
5553
Gene Name
proteoglycan 2, pro eosinophil major basic protein
Gene Aliases
BMPG, MBP, MBP1, proMBP
Chromosome Location
Chr.11: 57386787 - 57390657 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA Z26248.1 - 9 - 10 918 137

View Full Product Details