The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014096.3 | NP_054815.2 | 2 - 3 | 265 | 103 |
NM_017611.2 | NP_060081.1 | 2 - 3 | 101 | 103 | |
NM_199329.2 | NP_955361.1 | 2 - 3 | 150 | 103 | |
GenBank mRNA | AB028927.1 | - | 2 - 3 | 136 | 103 |
AK075552.1 | - | 2 - 3 | 123 | 103 | |
AK290391.1 | - | 1 - 2 | 163 | 103 | |
AK311401.1 | - | 2 - 3 | 120 | 103 | |
AL157431.1 | - | 2 - 3 | 170 | 103 | |
BC003163.1 | - | 2 - 3 | 102 | 103 | |
BG391025.1 | - | 2 - 3 | 227 | 102 | |
DC347054.1 | - | 2 - 3 | 150 | 103 | |
HY120731.1 | - | 2 - 3 | 265 | 103 |