The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290221.1 | NP_001277150.1 | 4 - 5 | 780 | 68 |
GenBank mRNA | AK056981.1 | - | 4 - 5 | 622 | 68 |