The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079673.1 | NP_001073141.1 | 4 - 5 | 552 | 65 |
NM_001278438.1 | NP_001265367.1 | 4 - 5 | 595 | 65 | |
NM_014923.4 | NP_055738.3 | 2 - 3 | 176 | 65 | |
NR_103528.1 | 4 - 5 | 595 | 65 | ||
XM_011534997.2 | XP_011533299.1 | 3 - 4 | 266 | 65 | |
XM_017020440.1 | XP_016875929.1 | 4 - 5 | 437 | 65 | |
XM_017020441.1 | XP_016875930.1 | 6 - 7 | 1058 | 65 | |
XM_017020442.1 | XP_016875931.1 | 5 - 6 | 590 | 65 | |
XM_017020443.1 | XP_016875932.1 | 4 - 5 | 910 | 65 | |
GenBank mRNA | AB023187.2 | - | 2 - 3 | 106 | 65 |
AJ749706.1 | - | 4 - 5 | 554 | 65 | |
AJ749707.1 | - | 2 - 3 | 167 | 65 | |
AK056071.1 | - | 4 - 5 | 536 | 65 | |
AK302415.1 | - | 2 - 3 | 176 | 65 | |
BC107793.1 | - | 2 - 3 | 97 | 65 | |
BC132812.1 | - | 4 - 5 | 319 | 65 | |
BC136617.1 | - | 4 - 5 | 320 | 65 | |
BC144301.1 | - | 4 - 5 | 320 | 65 | |
BX640739.1 | - | 4 - 5 | 455 | 65 | |
BX648141.1 | - | 4 - 5 | 542 | 65 | |
BX648706.1 | - | 4 - 5 | 550 | 65 |