The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079673.1 | NP_001073141.1 | 5 - 6 | 797 | 134 |
NM_001278438.1 | NP_001265367.1 | 5 - 6 | 840 | 134 | |
NM_014923.4 | NP_055738.3 | 3 - 4 | 421 | 134 | |
NR_103528.1 | 5 - 6 | 840 | 134 | ||
XM_011534997.2 | XP_011533299.1 | 4 - 5 | 511 | 134 | |
XM_017020440.1 | XP_016875929.1 | 5 - 6 | 682 | 134 | |
XM_017020441.1 | XP_016875930.1 | 7 - 8 | 1303 | 134 | |
XM_017020442.1 | XP_016875931.1 | 6 - 7 | 835 | 134 | |
XM_017020443.1 | XP_016875932.1 | 5 - 6 | 1155 | 134 | |
GenBank mRNA | AB023187.2 | - | 3 - 4 | 351 | 134 |
AJ749706.1 | - | 5 - 6 | 799 | 134 | |
AJ749707.1 | - | 3 - 4 | 412 | 134 | |
AK056071.1 | - | 5 - 6 | 781 | 134 | |
AK302415.1 | - | 3 - 4 | 421 | 134 | |
BC036010.1 | - | 1 - 2 | 217 | 134 | |
BC107793.1 | - | 3 - 4 | 342 | 134 | |
BC132812.1 | - | 5 - 6 | 564 | 134 | |
BC136617.1 | - | 5 - 6 | 565 | 134 | |
BC144301.1 | - | 5 - 6 | 565 | 134 | |
BX640739.1 | - | 5 - 6 | 700 | 134 | |
BX648141.1 | - | 5 - 6 | 787 | 134 |