The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079673.1 | NP_001073141.1 | 11 - 12 | 1530 | 96 |
NM_001278438.1 | NP_001265367.1 | 11 - 12 | 1573 | 96 | |
NM_014923.4 | NP_055738.3 | 9 - 10 | 1154 | 96 | |
NR_103528.1 | 11 - 12 | 1573 | 96 | ||
XM_011534997.2 | XP_011533299.1 | 10 - 11 | 1244 | 96 | |
XM_017020440.1 | XP_016875929.1 | 11 - 12 | 1415 | 96 | |
XM_017020441.1 | XP_016875930.1 | 13 - 14 | 2036 | 96 | |
XM_017020442.1 | XP_016875931.1 | 12 - 13 | 1568 | 96 | |
XM_017020443.1 | XP_016875932.1 | 11 - 12 | 1888 | 96 | |
GenBank mRNA | AB023187.2 | - | 9 - 10 | 1084 | 96 |
AJ749706.1 | - | 11 - 12 | 1532 | 96 | |
AJ749707.1 | - | 9 - 10 | 1145 | 96 | |
AK302415.1 | - | 9 - 10 | 1154 | 96 | |
BC060816.1 | - | 2 - 3 | 192 | 96 | |
BC070072.1 | - | 2 - 3 | 192 | 96 | |
BC132812.1 | - | 11 - 12 | 1297 | 96 | |
BC136617.1 | - | 11 - 12 | 1298 | 96 | |
BC144301.1 | - | 11 - 12 | 1298 | 96 | |
BX648141.1 | - | 11 - 12 | 1520 | 96 |