Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP11
Entrez Gene ID
8446
Gene Name
dual specificity phosphatase 11
Gene Aliases
PIR1
Chromosome Location
Chr.2: 73762184 - 73780157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003584.2 NP_003575.2 6 - 7 716 93
GenBank mRNA AF023917.1 - 6 - 7 657 93
AK315271.1 - 6 - 7 668 93

View Full Product Details