The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207362.2 | NP_997245.2 | 4 - 5 | 660 | 82 |
XM_005263923.3 | XP_005263980.1 | 4 - 5 | 640 | 82 | |
XM_005263924.3 | XP_005263981.1 | 4 - 5 | 640 | 82 | |
XM_005263926.3 | XP_005263983.1 | 4 - 5 | 525 | 82 | |
XM_011511093.1 | XP_011509395.1 | 6 - 7 | 799 | 82 | |
XM_011511094.1 | XP_011509396.1 | 5 - 6 | 697 | 82 | |
XM_011511095.2 | XP_011509397.1 | 4 - 5 | 467 | 82 | |
XM_017003971.1 | XP_016859460.1 | 4 - 5 | 643 | 82 | |
XM_017003972.1 | XP_016859461.1 | 5 - 6 | 747 | 82 | |
XM_017003973.1 | XP_016859462.1 | 4 - 5 | 653 | 82 | |
GenBank mRNA | BM922097.1 | - | 4 - 5 | 678 | 82 |