Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCO4
Entrez Gene ID
255104
Gene Name
transmembrane and coiled-coil domains 4
Gene Aliases
-
Chromosome Location
Chr.1: 19682213 - 19800385 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181719.4 NP_859070.3 15 - 16 1748 73
XM_005245822.4 XP_005245879.1 15 - 16 1764 73
XM_005245824.4 XP_005245881.1 15 - 16 1828 73
XM_005245825.4 XP_005245882.1 14 - 15 1629 73
XM_005245827.3 XP_005245884.1 14 - 15 1644 73
XM_006710551.3 XP_006710614.1 15 - 16 1853 73
XM_011541179.2 XP_011539481.1 18 - 19 2223 73
XM_011541180.2 XP_011539482.1 18 - 19 2378 73
XM_011541181.1 XP_011539483.1 17 - 18 2052 73
XM_011541185.2 XP_011539487.1 15 - 16 1771 73
XM_011541186.2 XP_011539488.1 15 - 16 1868 73
XM_017000913.1 XP_016856402.1 14 - 15 1652 73
XM_017000914.1 XP_016856403.1 17 - 18 2272 73
XM_017000915.1 XP_016856404.1 17 - 18 1994 73
XM_017000916.1 XP_016856405.1 15 - 16 1906 73
XM_017000917.1 XP_016856406.1 16 - 17 2121 73
XM_017000918.1 XP_016856407.1 17 - 18 2318 73
XM_017000919.1 XP_016856408.1 16 - 17 2045 73
XM_017000920.1 XP_016856409.1 15 - 16 2263 73
XM_017000921.1 XP_016856410.1 16 - 17 2070 73
XM_017000922.1 XP_016856411.1 14 - 15 1729 73
GenBank mRNA AK127543.1 - 4 - 5 525 73
AL832390.1 - 3 - 4 259 73
AX775875.1 - 14 - 15 1777 73
AX775877.1 - 13 - 14 1657 73
BC053600.1 - 13 - 14 1607 73
BX640987.1 - 15 - 16 1749 73

View Full Product Details