Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TMCO4
- Entrez Gene ID
-
255104
- Gene Name
- transmembrane and coiled-coil domains 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 19682213 - 19800385 on Build GRCh38
View Full Product Details